Merck
  • Dihydroartemisinin inhibits the tumorigenesis and metastasis of breast cancer via downregulating CIZ1 expression associated with TGF-β1 signaling.
Dihydroartemisinin inhibits the tumorigenesis and metastasis of breast cancer via downregulating CIZ1 expression associated with TGF-β1 signaling.
Life sciences (2020-02-24)
Yue Li, Xiaoyan Zhou, Jiali Liu, Ning Gao, Ruihua Yang, Qi Wang, Jing Ji, Ling Ma, Qian He
ABSTRACT
Dihydroartemisinin (DHA) is currently considered as the promising cancer therapeutic drug. In this study, we aimed to investigate the anti-proliferative and anti-metastasis effects of DHA. Utilizing breast cancer cells MCF-7, MDA-MB-231 and BT549, cell proliferation, migration and invasion were detected. RT-qPCR was performed to detect CIZ1, TGF-β1 and Snail expression, and the interactions of these related molecules were analyzed by GeneMANIA database. Western blot detected CIZ1, TGF-β1/Smads signaling and Snail expression in DHA-treated cells, in TGFβ1-induced cells with enhanced metastatic capacity, and in cells treated with DHA plus TGFβ1/TGFβ1 inhibitor SD-208. Results indicated DHA inhibited breast cancer cell proliferation and migration, with more potent effects compared with that of artemisinin. RT-qPCR and Western blot showed DHA inhibited CIZ1, TGF-β1 and Snail expression, and these molecules were shown to have protein-protein interactions by bioinformatics. Furthermore, TGFβ1-treatment enhanced MCF-7 migration and invasion, and CIZ1, TGF-β1/Smads signaling and snail activities; DHA, SD-208, combination of DHA and SD-208 reversed these conditions, preliminarily proving the cascade regulation between TGF-β1 signaling and CIZ1. MCF-7 xenografts model demonstrated the inhibition of DHA on tumor burden, and its mechanisms and well-tolerance in vivo; combination of DHA and SD-208 tried by us for the first time showed better treatment effects, but possible liver impairment made its use still keep cautious. DHA treatment inhibits the proliferation and metastasis of breast cancer, through suppressing TGF-β1/Smad signaling and CIZ1, suggesting the promising potential of DHA as a well-tolerated antitumor TGF-β1 pathway inhibitor.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder