Merck
  • Surface Acoustic Wave (SAW)-Enhanced Chemical Functionalization of Gold Films.
Surface Acoustic Wave (SAW)-Enhanced Chemical Functionalization of Gold Films.
Sensors (Basel, Switzerland) (2017-10-27)
Gina Greco, Matteo Agostini, Richie Shilton, Marco Travagliati, Giovanni Signore, Marco Cecchini
ABSTRACT
Surface chemical and biochemical functionalization is a fundamental process that is widely applied in many fields to add new functions, features, or capabilities to a material's surface. Here, we demonstrate that surface acoustic waves (SAWs) can enhance the chemical functionalization of gold films. This is shown by using an integrated biochip composed by a microfluidic channel coupled to a surface plasmon resonance (SPR) readout system and by monitoring the adhesion of biotin-thiol on the gold SPR areas in different conditions. In the case of SAW-induced streaming, the functionalization efficiency is improved ≈ 5 times with respect to the case without SAWs. The technology here proposed can be easily applied to a wide variety of biological systems (e.g., proteins, nucleic acids) and devices (e.g., sensors, devices for cell cultures).
Sigma-Aldrich
Latex beads, carboxylate-modified polystyrene, fluorescent red, aqueous suspension, 0.5 μm mean particle size
Pricing and availability is not currently available.