Merck
  • Simple means to alleviate sensitivity loss by trifluoroacetic acid (TFA) mobile phases in the hydrophilic interaction chromatography?electrospray tandem mass spectrometric (HILIC?ESI/MS/MS) bioanalysis of basic compounds
Simple means to alleviate sensitivity loss by trifluoroacetic acid (TFA) mobile phases in the hydrophilic interaction chromatography?electrospray tandem mass spectrometric (HILIC?ESI/MS/MS) bioanalysis of basic compounds
Journal of Chromatography. B, Biomedical Sciences and Applications (2005)
Shou ZW and Naidong W