Merck
  • Does bone scintigraphy still have a role in the era of 68 Ga-PSMA PET/CT in prostate cancer?
Does bone scintigraphy still have a role in the era of 68 Ga-PSMA PET/CT in prostate cancer?
Annals of nuclear medicine (2020-05-13)
Duygu Has Simsek, Yasemin Sanli, Caner Civan, Muge Nur Engin, Emine Goknur Isik, Zeynep Gozde Ozkan, Serkan Kuyumcu
ABSTRACT
We aimed to evaluate the role of bone scintigraphy (BS) which has long been the imaging modality of choice in prostate cancer (PCa) and performed a head-to-head comparison between BS, BS + SPECT/CT and 68 Ga-PSMA-PET/CT, for the detection of bone metastasis of PCa. We evaluated 138 PCa patients who underwent BS and 68 Ga-PSMA PET/CT and SPECT/CT of 102 of 138 patients. Images were interpreted retrospectively and areas of abnormally increased tracer uptake related to PCa were documented as benign, metastatic or equivocal. Equivocal uptakes were finally diagnosed based on a consensus review of correlative imaging. Patient- and lesion-based analysis was performed. Patients with superscan images were excluded from lesion-based analysis. At least one metastatic or equivocal uptake in skeleton was defined in 76 of 138 (55%) BS, in 33 of 102 (32.3%) SPECT/CT, and in 49 of 138 (35.5%) 68 Ga-PSMA PET/CT. 23 (16.7%) patients had also superscan findings on BS and 68 Ga-PSMA PET/CT. For patient-based analysis, sensitivity, specificity, accuracy, PPV, and NPV were calculated as 91.1%, 64.5%, 73.1%, 55.4% and 93.7% for BS; 95.5%, 82.7%, 86.9%, 72.8% and 96.2% for BS + SPECT/CT; 97.7%, 95.7%, 95.6%, 91.6% and 98.8% for 68 Ga-PSMA PET/CT. For lesion-based analysis, sensitivity, specificity, accuracy, PPV, and NPV were 53%, 63.9%, 60.1%, 42.8% and 71.8% for BS; 59.2%, 87.6%, 77.7%, 62.6% and 80% for BS + SPECT/CT; 96.4%, 98.1%, 97.5%, 96.4% and 98.1% for 68 Ga-PSMA PET/CT. This study has shown that 68 Ga-PSMA PET/CT overcomes the limitations of BS and proves superiority in detecting bone metastases, even in patients with SPECT/CT. Our findings present important implications that 68 Ga-PSMA PET/CT can replace BS in future practice.
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%