Merck
  • Oral delivery of maize-produced porcine epidemic diarrhea virus spike protein elicits neutralizing antibodies in pigs.
Oral delivery of maize-produced porcine epidemic diarrhea virus spike protein elicits neutralizing antibodies in pigs.
Plant cell, tissue and organ culture (2020-05-13)
Erin Egelkrout, Celine Hayden, Gina Fake, Todd Keener, Paulo Arruda, Ryan Saltzman, John Walker, John Howard
ABSTRACT
Porcine Epidemic Diarrhea Virus (PEDV) causes severe diarrhea and mortality in piglets. Robust immunity may break the transmission cycle. Expression of antigens in maize grains is a promising method for producing low-cost vaccines. As a first step, we expressed maize constructs containing PEDV S1 spike protein targeted to various cellular locations including the cell wall, endoplasmic reticulum, and vacuole, and fused to carrier proteins E. coli heat labile subunit (LTB) and a dendritic cell (DC) binding peptide, and obtained sufficient antigen for oral immunization. Constructs targeting S1 to the ER or fused to carrier proteins produced high levels of antigen of greater than 20 mg/kg. Oral administration to pigs elicited serum neutralizing antibodies, supporting oral immunization as a practical and cost-effective PEDV vaccine.
Sigma-Aldrich
Pyridinium dichromate, 98%
Sigma-Aldrich
BCIP®/NBT Liquid Substrate System, ready to use solution