Merck
  • Ortho and para oxydehalogenation of dihalophenols catalyzed by the monooxygenase TcpA and NAD(P)H:FAD reductase Fre.
Ortho and para oxydehalogenation of dihalophenols catalyzed by the monooxygenase TcpA and NAD(P)H:FAD reductase Fre.
Journal of hazardous materials (2019-12-11)
Liancheng Fang, Han Qin, Taozhong Shi, Xiangwei Wu, Qing X Li, Rimao Hua
ABSTRACT
Dihalophenols such as dichlorophenols (DCPs) are important industrial chemical intermediates, but also persistent pollutants in the environment. Oxidative dehalogenation by microbes is an efficient biological method to degrade halophenols, but the mechanism is unclear yet. Cupriavidus nantongensis X1T was a type strain of genus Cupriavidus, and could degrade 2,4-dichlorophenol of 50 mg/L within 12 h. The degradation rate constant was approximately 84 fold greater than that by Bacillus endophyticus CP1R43, a well-studied 2,4-DCP-degrading bacterial strain. The genes encoding 2,4,6-trichlorophenol monooxygenase (TcpA) and NAD(P)H:FAD reductase (Fre) from strain X1T were cloned and expressed. The expressed TcpA Fre were purified. The molecular docking of TcpA with DCPs and point mutation experiments showed that the degradation activity of TcpA was associated with the length of the hydrogen bond between the substrates and the amino acids in the active pocket. DCPs were degraded via a stepwise oxidative dechlorination in a positive relationship between the oxidation ability and the electron-withdrawing potential of the p-position group. In addition, TcpA has dual dehalogenation and denitration functions. The results demonstrate that either strain X1T or TcpA and Fre can effectively dehalogenate dihalophenols, which can be useful for the treatment of dihalophenols in wastewaters and remediation of DCP-contaminated environments.
Sigma-Aldrich
3-Chlorophenol, 98%
Sigma-Aldrich
2-Chloro-1,4-benzoquinone, 95%
Sigma-Aldrich
4-Bromo-2-chlorophenol, 99%
Sigma-Aldrich
Chlorohydroquinone, technical grade, 85%