Merck
  • Illuminating NAD+ Metabolism in Live Cells and In Vivo Using a Genetically Encoded Fluorescent Sensor.
Illuminating NAD+ Metabolism in Live Cells and In Vivo Using a Genetically Encoded Fluorescent Sensor.
Developmental cell (2020-03-21)
Yejun Zou, Aoxue Wang, Li Huang, Xudong Zhu, Qingxun Hu, Yinan Zhang, Xianjun Chen, Fengwen Li, Qiaohui Wang, Hu Wang, Renmei Liu, Fangting Zuo, Ting Li, Jing Yao, Yajie Qian, Mei Shi, Xiao Yue, Weicai Chen, Zhuo Zhang, Congrong Wang, Yong Zhou, Linyong Zhu, Zhenyu Ju, Joseph Loscalzo, Yi Yang, Yuzheng Zhao
ABSTRACT
Understanding of NAD+ metabolism provides many critical insights into health and diseases, yet highly sensitive and specific detection of NAD+ metabolism in live cells and in vivo remains difficult. Here, we present ratiometric, highly responsive genetically encoded fluorescent indicators, FiNad, for monitoring NAD+ dynamics in living cells and animals. FiNad sensors cover physiologically relevant NAD+ concentrations and sensitively respond to increases and decreases in NAD+. Utilizing FiNad, we performed a head-to-head comparison study of common NAD+ precursors in various organisms and mapped their biochemical roles in enhancing NAD+ levels. Moreover, we showed that increased NAD+ synthesis controls morphofunctional changes of activated macrophages, and directly imaged NAD+ declines during aging in situ. The broad utility of the FiNad sensors will expand our mechanistic understanding of numerous NAD+-associated physiological and pathological processes and facilitate screening for drug or gene candidates that affect uptake, efflux, and metabolism of this important cofactor.
Sigma-Aldrich
FK866 hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Sodium oxamate, ≥98%
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type II, ammonium sulfate suspension, 350-600 units/mg protein
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder