Merck
  • Enantioanalysis of glutamine-a key factor in establishing the metabolomics process in gastric cancer.
Enantioanalysis of glutamine-a key factor in establishing the metabolomics process in gastric cancer.
Analytical and bioanalytical chemistry (2020-03-19)
Raluca-Ioana Stefan-van Staden, Ruxandra-Maria Ilie-Mihai, Lidia Magerusan, Maria Coros, Stela Pruneanu
ABSTRACT
Gastric cancer is the second leading cause of death in the world. Early detection will facilitate early treatment and full recovery of the patients. Metabolomics facilitated the detection of few amino acids able to be used as biomarkers. It was always assumed that the L-enantiomer of the chiral amino acid is part of the process, forgetting the twisting of DNA molecules which may also produce the D-enantiomer of the amino acid. Therefore, an enantioanalysis of amino acids such as glutamine which are part of the gastric cancer metabolomics is absolutely necessary. Four stochastic sensors based on immobilization of protoporphyrin IX, β-cyclodextrin, and 2,2-diphenyl-1-picrylhydrazyl on graphene materials were proposed for the enantioanalysis of glutamine in whole blood samples of patients with gastric cancer. Different signatures were obtained for the enantiomers for each stochastic sensor, making possible the enantioanalysis of glutamine in large concentration ranges-from fmol/L to mmol/L; these ranges facilitating the enantioanalysis of glutamine in healthy patients as well as in patients were found in early and later stages of gastric cancer. Graphical abstract.
Sigma-Aldrich
D-Glutamine, ≥98% (HPLC)