Merck
seco-iridoids from Calycophyllum spruceanum (Rubiaceae).
Phytochemistry (2003-08-29)
Luz Margarita Cardona Zuleta, Alberto José Cavalheiro, Dulce Helena Siqueira Silva, Maysa Furlan, Maria Claudia Marx Young, Sérgio Albuquerque, Ian Castro-Gamboa, Vanderlan da Silva Bolzani
ABSTRACT
Three seco-iridoids 7-methoxydiderroside, 6'-O-acetyldiderroside and 8-O-tigloyldiderroside, were isolated from the wood bark of Calycophyllum spruceanum together with the known iridoids loganetin, loganin and the seco-iridoids secoxyloganin, kingiside and diderroside. Their structures were elucidated by means of NMR and MS spectral data analysis. Using NOE correlations and coupling constants, the relative stereochemistry of the new derivatives was established. 7-Methoxydiderroside, 6'-O-acetyldiderroside and the known secoxyloganin and diderroside showed in vitro activity against trypomastigote forms of Trypanosoma cruzi, with IC(50) values of 59.0, 90.2, 74,2 and 84.9 microg/mL, respectively and were compared to the standard gentian violet (IC(50) 7.5 microg/ml).
Sigma-Aldrich
Diderroside, ≥90% (LC/MS-ELSD)