Merck
  • Chicken avian β-defensin 8 modulates immune response via the mitogen-activated protein kinase signaling pathways in a chicken macrophage cell line.
Chicken avian β-defensin 8 modulates immune response via the mitogen-activated protein kinase signaling pathways in a chicken macrophage cell line.
Poultry science (2020-09-02)
Yeojin Hong, Jiae Lee, Thi Hao Vu, Sooyeon Lee, Hyun S Lillehoj, Yeong Ho Hong
ABSTRACT
Defensins are antimicrobial peptides composed of 3 conserved disulfide bridges, a β-sheet, and both hydrophobic and cationic amino acids. In this study, we aimed to demonstrate the immunomodulation role of avian β-defensin 8 (AvBD8) in a chicken macrophage cell line. Chicken AvBD8 stimulated the expression of proinflammatory cytokines (IL-1β, interferon gamma, and IL-12p40) and chemokines (CCL4, CXCL13, and CCL20) in macrophages. Furthermore, by Western blotting and immunocytochemistry, we confirmed that AvBD8 activated the mitogen-activated protein kinase signaling pathway via extracellular regulated kinases 1/2 and p38 signaling molecules. Overall, AvBD8 plays a crucial role in host defense as not only an antimicrobial peptide but also an immunomodulator by activating the mitogen-activated protein kinase signaling pathway and inducing the expression of proinflammatory cytokines and chemokines.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O