Merck
  • Intercalated disc protein Xinβ is required for Hippo-YAP signaling in the heart.
Intercalated disc protein Xinβ is required for Hippo-YAP signaling in the heart.
Nature communications (2020-09-18)
Haipeng Guo, Yao Wei Lu, Zhiqiang Lin, Zhan-Peng Huang, Jianming Liu, Yi Wang, Hee Young Seok, Xiaoyun Hu, Qing Ma, Kathryn Li, Jan Kyselovic, Qingchuan Wang, Jenny L-C Lin, Jim J-C Lin, Douglas B Cowan, Francisco Naya, Yuguo Chen, William T Pu, Da-Zhi Wang
ABSTRACT
Intercalated discs (ICD), specific cell-to-cell contacts that connect adjacent cardiomyocytes, ensure mechanical and electrochemical coupling during contraction of the heart. Mutations in genes encoding ICD components are linked to cardiovascular diseases. Here, we show that loss of Xinβ, a newly-identified component of ICDs, results in cardiomyocyte proliferation defects and cardiomyopathy. We uncovered a role for Xinβ in signaling via the Hippo-YAP pathway by recruiting NF2 to the ICD to modulate cardiac function. In Xinβ mutant hearts levels of phosphorylated NF2 are substantially reduced, suggesting an impairment of Hippo-YAP signaling. Cardiac-specific overexpression of YAP rescues cardiac defects in Xinβ knock-out mice-indicating a functional and genetic interaction between Xinβ and YAP. Our study reveals a molecular mechanism by which cardiac-expressed intercalated disc protein Xinβ modulates Hippo-YAP signaling to control heart development and cardiac function in a tissue specific manner. Consequently, this pathway may represent a therapeutic target for the treatment of cardiovascular diseases.
Sigma-Aldrich
MISSION® esiRNA, targeting human XIRP2
Sigma-Aldrich
ApopTag Fluorescein In Situ Apoptosis Detection Kit, The ApopTag Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a Fluorescein reporter molecule.