Merck
  • Non-Arsenical heartworm adulticidal therapy using topical moxidectin-imidacloprid and doxycycline: A prospective case series.
Non-Arsenical heartworm adulticidal therapy using topical moxidectin-imidacloprid and doxycycline: A prospective case series.
Veterinary parasitology (2020-05-26)
Marisa K Ames, Philip VanVranken, Chelsea Evans, Clarke E Atkins
ABSTRACT
This prospective case series evaluated the adulticidal efficacy of topical 10 % moxidectin/2.5 % imidacloprid (M/I; Advantage Multi®, Bayer, Shawnee Mission, KS, USA) and doxycycline in dogs with naturally occurring heartworm infection (HWI). Twenty-two dogs with HWI whose owners declined melarsomine were treated with M/I at the preventive dosage twice monthly for 90 days then monthly thereafter and doxycycline (median [interquartile range; IQR] dosage 12.6 [12.0-16.1] mg/kg/day) for the first 15 days. Although strict activity restriction was not imposed, owners were asked to prevent their dogs from exercising strenuously. This protocol was referred to as the MOXY protocol. Antigen testing was performed every 30-60 days, until dogs had 'no antigen detected' (NAD). Twenty-one of the 22 dogs ultimately converted to NAD by 434 days (median [IQR]), 234 (179-303). One dog remained positive 701 days after MOXY initiation and was considered a treatment failure. All sera which converted to NAD on HW antigen testing were retested after heat-treatment. Twelve dogs had NAD on the heat-treated test on the same day as having their first NAD on the conventional test. Six of 9 dogs testing positive after heat-treatment were retested and all 6 had NAD on a heat-treated test within 2-3 months. Microfilaremia was cleared in all 8 dogs re-tested. Four dogs required treatment for cough, thought due to heartworm (HW) death, an average of 89 days after initiation of MOXY. This cough was most likely due to pneumonitis with heartworm-pulmonary thromboembolism. One dog required hospitalization for 24 -h and recovered fully with corticosteroid therapy and supportive care and 2 dogs were treated in an outpatient fashion with steroids. The MOXY protocol was tolerated and 96 % (21/22) of dogs converted to NAD, though 2 dogs required greater than 1 year to achieve this result. Nonaresenical-adulticide therapy may result in pneumonitis and heartworm-pulmonary thromboembolism at unpredictable times, potentially months after initiation of macrocyclic lactone therapy and exercise restriction should be considered when using a nonarsenical protocol. Although not currently recommended by the American Heartworm Society (AHS), non-arsenical strategies are in use and the goal of this study was to evaluate the efficacy, duration of therapy, and safety of an accelerated dosing protocol of M/I with doxycycline.
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder