Merck
  • Distinct contributions of two choline-producing enzymatic activities to lysophosphatidic acid production in human amniotic fluid from pregnant women in the second trimester and after parturition.
Distinct contributions of two choline-producing enzymatic activities to lysophosphatidic acid production in human amniotic fluid from pregnant women in the second trimester and after parturition.
Prostaglandins & other lipid mediators (2020-06-26)
Midori Fukui, Toshihiko Tsutsumi, Aimi Yamamoto-Mikami, Katsuya Morito, NaokoTakahashi, Tamotsu Tanaka, Takeshi Iwasa, Akira Kuwahara, Minoru Irahara, Akira Tokumura
ABSTRACT
The purpose of this study was to clarify whether human amniotic fluid (AF) contains a significant level of bioactive lysophosphatidic acid (LPA) and, whether autotaxin (ATX) is involved in the production of LPA, if present. Using LC-MS/MS, we found a higher ratio of levels of LPA and its precursor lysophosphatidylcholine (LPC) in AF collected after parturition than that in AF collected at the middle stage of pregnancy. We detected significant choline-producing enzymatic activity toward an exogenous LPC in AF at the middle stage of pregnancy, about half of which was ascribable to ATX. In AF collected after parturition, the ATX-independent choline-producing activity of glycerophosphcholine phosphodiesterase coupled to lysophospholipase A activity was increased in relative to the lysophospholipase D activity of ATX. These results suggest that the increased LPA/LPC ratio in AF at the term of pregnancy was due to not only a moderate increase in the level of LPC, but also an unknown mechanism involving epithelial cells bathed with AF.
Sigma-Aldrich
Phosphocholine chloride calcium salt tetrahydrate, Sigma Grade
Pricing and availability is not currently available.