Merck
  • Expression, epitope prediction and IgE-binding of the Tyrophagus putrescentiae group 13 allergen.
Expression, epitope prediction and IgE-binding of the Tyrophagus putrescentiae group 13 allergen.
Experimental and therapeutic medicine (2020-09-17)
Nan Wang, Ying Zhou, Meili Wu, Hanting Zhu, Yubao Cui
ABSTRACT
Storage mites, such as Tyrophagus putrescentiae, are an important source of allergens that cause allergic diseases in humans. It has previously been indicated that T. putrescentiae has a high sensitization rate as an allergen in some Asian and European countries. Identifying and cloning the allergens in this species may enable improved diagnostic and therapeutic approaches. The aim of the present study was to clone and sequence the T. putrescentiae group 13 allergen (Tyr p 13) isolated from storage mites in China, to use bioinformatics tools to model its biophysical characteristics and to induce protein expression to test its IgE-binding activity. The full-length cDNA comprised 486 bp and was predicted to include a signal peptide of 22 amino acids. Its secondary structure was shown to comprise an α-helix (10.79%), extended strand (33.81%) and random coils (55.40%). Using homology modeling, the present study constructed a reasonable tertiary structure of Tyr p 13. Linear Bcell epitopes at amino acids 47-53, 70-76, 81-86, 101-105 and 112120 were predicted. Three discontinuous B-cell epitopes were also predicted: i) 47, 48, 49, 50, 51, 52, 53, 70, 71, 72 and 73; ii) 91, 92, 93, 94, 112, 113, 114, 115, 116, 117, 118, 119, 120, 121 and 138; and iii) 74, 76, 79, 81, 82, 83, 84, 86, 101, 102, 103, 104 and 105. SDS-PAGE identified a specific band at the predicted molecular weight of the recombinant Tyr p 13 (rTyr p 13), demonstrating its successful expression. The rTyr p 13 bound to IgE in the serum of 13.2% (5/38) of patients allergic to T. putrescentiae, according to ELISA. The successful cloning of Tyr p 13 and basic bioinformatics analysis of the protein provided a foundation for the further study of this allergen with regards to the diagnosis and treatment of patients allergic to storage mites. These results provided a theoretical basis for the design of rTyr p 13 with modified B-cell epitopes.
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder