Merck
  • Occurrence and emission of phthalates, bisphenol A, and oestrogenic compounds in concentrated animal feeding operations in Southern China.
Occurrence and emission of phthalates, bisphenol A, and oestrogenic compounds in concentrated animal feeding operations in Southern China.
Ecotoxicology and environmental safety (2020-12-02)
Huan-Yu Tao, Jiawei Zhang, Jianghong Shi, Wei Guo, Xiaowei Liu, Mengtao Zhang, Hui Ge, Xiao-Yan Li
ABSTRACT
Phthalates (PAEs), bisphenol A (BPA), and oestrogenic compounds have become major concerns due to their endocrine-disrupting effect. However, few studies related to the occurrence of PAEs, BPA, and oestrogen in food and compost from different growth age livestock have been conducted. In this study, faeces, urine and food samples were collected from a typical livestock (cow) and a special livestock (pigeon) from concentrated animal feeding operations (CAFOs). The daily total oestrogen excretion of a single cow ranged from 192 μg/day to 671 μg/day, which was significantly higher than that of a single pigeon (0-0.01 μg/day). Conjugated oestrogens represented 22.0-46.0% of the total oestrogens excreted from cow faeces and 80.7-91.8% of those from cow urine, indicating that the form of the excreted oestrogens depends on the livestock species and type of excrement. BPA was all detected in all livestock manure and food, and the concentration in pigeon was 9.2-40.2 ng/g and 23.1 ng/g respectively, while that in cattle was 50.5-72.0 ng/g and 41.1 ng/g respectively. The results indicated that the food is significant sources of BPA entering the process of cow and pigeon breeding. Diethyl phthalate (DEP) was detected at high frequency in pigeon faeces samples, suggesting that pigeons were highly exposed to these plasticisers. The total oestradiol equivalent quantity (EEQt) of livestock origin in aquatic environments was estimated to be 2.99 ng/L, which was higher than the baseline hazard value (1 ng/L) (Xu et al., 2018). The study provides data on the emissions and sources of PAEs, BPA, and oestrogenic compounds from different livestock in CAFOs and demonstrates that food is a significant source of BPA entering livestock.
Sigma-Aldrich
Dess-Martin periodinane, 97%