Merck
  • Astrocytes are necessary for blood-brain barrier maintenance in the adult mouse brain.
Astrocytes are necessary for blood-brain barrier maintenance in the adult mouse brain.
Glia (2020-09-22)
Benjamin P Heithoff, Kijana K George, Aubrey N Phares, Ivan A Zuidhoek, Carmen Munoz-Ballester, Stefanie Robel
ABSTRACT
In the adult brain, multiple cell types are known to produce factors that regulate blood-brain barrier (BBB) properties, including astrocytes. Yet several recent studies disputed a role for mature astrocytes at the BBB. To determine if astrocytes contribute a nonredundant and necessary function in maintaining the adult BBB, we used a mouse model of tamoxifen-inducible astrocyte ablation. In adult mice, tamoxifen induction caused sparse apoptotic astrocyte cell death within 2 hr. Indicative of BBB damage, leakage of the small molecule Cadaverine, and the large plasma protein fibrinogen into the brain parenchyma indicative of BBB damage was detected as early as astrocyte ablation was present. Vessels within and close to regions of astrocyte loss had lower expression of the tight junction protein zonula occludens-1 while endothelial glucose transporter 1 expression was undisturbed. Cadaverine leakage persisted for several weeks suggesting a lack of barrier repair. This is consistent with the finding that ablated astrocytes were not replaced. Adjacent astrocytes responded with partial nonproliferative astrogliosis, characterized by morphological changes and delayed phosphorylation of STAT3, which restricted dye leakage to the brain and vessel surface areas lacking coverage by astrocytes 1 month after ablation. In conclusion, astrocytes are necessary to maintain BBB integrity in the adult brain. BBB-regulating factors secreted by other cell types, such as pericytes, are not sufficient to compensate for astrocyte loss.
Sigma-Aldrich
Anti-Water Channel Aquaporin 4 antibody produced in rabbit, lyophilized powder, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-Glutamate Transporter Antibody, Glial, serum, Chemicon®
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit