Merck
  • Novel Copper Photoredox Catalysts for Polymerization: An In Situ Synthesis of Metal Nanoparticles.
Novel Copper Photoredox Catalysts for Polymerization: An In Situ Synthesis of Metal Nanoparticles.
Polymers (2020-10-11)
Haja Tar, Tahani I Kashar, Noura Kouki, Reema Aldawas, Bernadette Graff, Jacques Lalevée
ABSTRACT
The copper II complex (HLCuCl) carrying 2,4 dinitrophenylhydrazone (L) is synthesized and evaluated as a new photoredox catalyst/photoinitiator in combination with triethylamine (TEA) and iodonium salt (Iod) for the radical polymerization of ethylene glycol diacrylate during exposure to visible light using a photoreactor at 419 nm. The copper complex reactivity with TEA/Iod salt/gold chloride showed a good production and stability of gold nanoparticles. Finally, the high performance of Cu (II) complex for radical photopolymerization incorporating gold nanoparticles is provided. The photochemical mechanisms for the production of initiating radicals are studied using cyclic voltammetry. Polymer nanocomposites containing gold nanoparticles (Au NPs) in situ photogenerated during the irradiation process were prepared. The formation of Au NPs inside the polymer matrix was through UV-Vis and EDS/SEM analyses.
Sigma-Aldrich
Diphenyliodonium hexafluorophosphate, ≥98%