Merck
  • Synthesis of indoleamine 2,3-dioxygenase inhibitory analogues of the sponge alkaloid exiguamine A.
Synthesis of indoleamine 2,3-dioxygenase inhibitory analogues of the sponge alkaloid exiguamine A.
Journal of medicinal chemistry (2008-04-09)
Gavin Carr, Marco K W Chung, A Grant Mauk, Raymond J Andersen
ABSTRACT
Synthetic analogues of the sponge natural product exiguamine A (3) have been prepared and evaluated for their ability to inhibit indoleamine 2,3-dioxygenase in vitro.
Sigma-Aldrich
4-Hydroxyindole, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Methyl-DL-tryptophan, 97%
Pricing and availability is not currently available.