Merck
  • Housing temperature affects the acute and chronic metabolic adaptations to exercise in mice.
Housing temperature affects the acute and chronic metabolic adaptations to exercise in mice.
The Journal of physiology (2019-07-13)
Greg L McKie, Kyle D Medak, Carly M Knuth, Hesham Shamshoum, Logan K Townsend, Willem T Peppler, David C Wright
ABSTRACT
Mice are commonly housed at room temperatures below their thermoneutral zone meaning they are exposed to chronic thermal stress. Endurance exercise induces browning and mitochondrial biogenesis in white adipose tissue of rodents, but there are conflicting reports of this phenomenon in humans. We hypothesized that the ambient room temperature at which mice are housed could partially explain these discrepant reports between humans and rodents. We housed mice at room temperature or thermoneutrality and studied their physiological responses to acute and chronic exercise. We found that thermoneutral housing altered running behaviour and glucose homeostasis, and further, that exercise-induced markers of mitochondrial biogenesis and the browning of white adipose tissue were reduced in mice housed at thermoneutrality. Mice are often housed at temperatures below their thermoneutral zone resulting in compensatory increases in thermogenesis. Despite this, many studies report housing mice at room temperature (RT), likely for the convenience of the researchers studying them. As such, the conflicting reports between humans and rodents regarding the ability of exercise to increase mitochondrial and thermogenic markers in white adipose tissue may be explained by the often-overlooked variable, housing temperature. To test this hypothesis, we housed male C57BL/6 mice at RT (22°C) or thermoneutrality (TN) (29°C) with or without access to a voluntary running wheel for 6 weeks or subjected them to an acute exhaustive bout of treadmill running. We examined the gene expression and protein content of select mitochondrial and thermogenic markers in skeletal muscle, epididymal white adipose tissue (eWAT), inguinal white adipose tissue (iWAT) and brown adipose tissue (BAT). We also assessed adipocyte morphology and indices of glucose homeostasis. Housing temperature influenced glucose tolerance and insulin action in vivo, yet the beneficial effects of exercise, both acute and chronic, remained intact in eWAT, BAT and skeletal muscle irrespective of housing temperature. Housing mice at TN led to an attenuation of some of the effects of exercise on iWAT. Collectively, we present data characterizing the acute and chronic metabolic adaptations to exercise at different housing temperatures and demonstrate, for the first time, that temperature influences the ability of exercise to increase markers of mitochondrial biogenesis and the browning of white adipose tissue.
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, >= 550 units/mg protein (biuret)
Sigma-Aldrich
Anti-PGC-1 Antibody, Chemicon®, from rabbit