Merck
  • Lignans from the stems and leaves of Brandisia hancei and their effects on VEGF-induced vascular permeability and migration of HRECs and DLAV formation in zebrafish.
Lignans from the stems and leaves of Brandisia hancei and their effects on VEGF-induced vascular permeability and migration of HRECs and DLAV formation in zebrafish.
Bioscience, biotechnology, and biochemistry (2014-12-17)
Ik-Soo Lee, Young Sook Kim, Seung-Hyun Jung, Song Yi Yu, Joo-Hwan Kim, Hang Sun, Jin Sook Kim
ABSTRACT
In our continuing search for novel antiangiogenic agents, a new lignan glycoside, (7R,8R)-1-(4-O-β-d-glucopyranosyl-3-methoxyphenyl)-2-{2-methoxy-4-[1-(E)-propene-3-ol]-phenoxyl}-propane-1,3-diol (1), along with three known lignans (2-4), were isolated from the 80% EtOH extract of Brandisia hancei stems and leaves. These isolates (1-4) were subjected to an in vitro bioassay to evaluate their effects on vascular endothelial growth factor (VEGF)-induced vascular permeability and migration of human retinal endothelial cells (HRECs). Of the compounds tested, compound 1 resulted in the greatest reduction in VEGF-induced vascular permeability by about 31.5% at 10 μM compared to the VEGF-treated control. In the migration assay, compounds 1 and 2 significantly decreased VEGF-induced HREC migration. Furthermore, zebrafish embryos treated with compounds 1 and 2 showed mild reductions of dorsal longitudinal anastomotic vessel (DLAV) formation.
Sigma-Aldrich
In Vitro Vascular Permeability Assay (24-well), This In Vitro Vascular Permeability Assay kit employs a 24-well plate and provides an efficient system for evaluating the effects of chemicals & drug compounds on endothelial cell adsorption, transport & permeability.
Pricing and availability is not currently available.