Merck
  • Synthesis, characterization, antioxidant activity and DNA-binding studies of two rare earth(III) complexes with naringenin-2-hydroxy benzoyl hydrazone ligand.
Synthesis, characterization, antioxidant activity and DNA-binding studies of two rare earth(III) complexes with naringenin-2-hydroxy benzoyl hydrazone ligand.
European journal of medicinal chemistry (2007-11-27)
Tian-Rong Li, Zheng-Yin Yang, Bao-Dui Wang, Dong-Dong Qin
ABSTRACT
Two novel rare earth complexes, Y(III) complex (1) and Eu(III) complex (2), with naringenin-2-hydroxy benzoyl hydrazone ligand were synthesized and characterized. The interaction of the two metal complexes and the free ligand with calf thymus DNA (CT DNA) was investigated by electronic absorption spectroscopy, fluorescence spectroscopy and viscosity measurement. All the experimental evidences indicate that these three compounds can strongly bind to CT DNA via an intercalation mechanism. The intrinsic binding constants of the Y(III) complex (1), Eu(III) complex (2) and the free ligand with CT DNA were 2.1 x 10(4), 8.5 x 10(4) and 1.6 x 10(4) M(-1), respectively. Furthermore, the antioxidant activity of the metal complexes was determined by hydroxyl radical scavenging method in vitro.
Product Description
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)