Merck
  • Cadmium induces autophagy through ROS-dependent activation of the LKB1-AMPK signaling in skin epidermal cells.
Cadmium induces autophagy through ROS-dependent activation of the LKB1-AMPK signaling in skin epidermal cells.
Toxicology and applied pharmacology (2011-07-20)
Young-Ok Son, Xin Wang, John Andrew Hitron, Zhuo Zhang, Senping Cheng, Amit Budhraja, Songze Ding, Jeong-Chae Lee, Xianglin Shi
ABSTRACT
Cadmium is a toxic heavy metal which is environmentally and occupationally relevant. The mechanisms underlying cadmium-induced autophagy are not yet completely understood. The present study shows that cadmium induces autophagy, as demonstrated by the increase of LC3-II formation and the GFP-LC3 puncta cells. The induction of autophagosomes was directly visualized by electron microscopy in cadmium-exposed skin epidermal cells. Blockage of LKB1 or AMPK by siRNA transfection suppressed cadmium-induced autophagy. Cadmium-induced autophagy was inhibited in dominant-negative AMPK-transfected cells, whereas it was accelerated in cells transfected with the constitutively active form of AMPK. mTOR signaling, a negative regulator of autophagy, was downregulated in cadmium-exposed cells. In addition, cadmium generated reactive oxygen species (ROS) at relatively low levels, and caused poly(ADP-ribose) polymerase-1 (PARP) activation and ATP depletion. Inhibition of PARP by pharmacological inhibitors or its siRNA transfection suppressed ATP reduction and autophagy in cadmium-exposed cells. Furthermore, cadmium-induced autophagy signaling was attenuated by either exogenous addition of catalase and superoxide dismutase, or by overexpression of these enzymes. Consequently, these results suggest that cadmium-mediated ROS generation causes PARP activation and energy depletion, and eventually induces autophagy through the activation of LKB1-AMPK signaling and the down-regulation of mTOR in skin epidermal cells.
Sigma-Aldrich
Anti-PAR (Ab-1) Mouse mAb (10H), liquid, ≥95% (SDS-PAGE), clone 10H
Pricing and availability is not currently available.