Merck
  • Broad and potent neutralizing human antibodies to tick-borne flaviviruses protect mice from disease.
Broad and potent neutralizing human antibodies to tick-borne flaviviruses protect mice from disease.
The Journal of experimental medicine (2021-04-09)
Marianna Agudelo, Martin Palus, Jennifer R Keeffe, Filippo Bianchini, Pavel Svoboda, Jiří Salát, Avery Peace, Anna Gazumyan, Melissa Cipolla, Tania Kapoor, Francesca Guidetti, Kai-Hui Yao, Jana Elsterová, Dana Teislerová, Aleš Chrdle, Václav Hönig, Thiago Oliveira, Anthony P West, Yu E Lee, Charles M Rice, Margaret R MacDonald, Pamela J Bjorkman, Daniel Růžek, Davide F Robbiani, Michel C Nussenzweig
ABSTRACT
Tick-borne encephalitis virus (TBEV) is an emerging human pathogen that causes potentially fatal disease with no specific treatment. Mouse monoclonal antibodies are protective against TBEV, but little is known about the human antibody response to infection. Here, we report on the human neutralizing antibody response to TBEV in a cohort of infected and vaccinated individuals. Expanded clones of memory B cells expressed closely related anti-envelope domain III (EDIII) antibodies in both groups of volunteers. However, the most potent neutralizing antibodies, with IC50s below 1 ng/ml, were found only in individuals who recovered from natural infection. These antibodies also neutralized other tick-borne flaviviruses, including Langat, louping ill, Omsk hemorrhagic fever, Kyasanur forest disease, and Powassan viruses. Structural analysis revealed a conserved epitope near the lateral ridge of EDIII adjoining the EDI-EDIII hinge region. Prophylactic or early therapeutic antibody administration was effective at low doses in mice that were lethally infected with TBEV.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, FITC conjugate, Species Adsorbed, 1.3 mg/mL, Chemicon®
Pricing and availability is not currently available.