Merck
  • The RNA helicase Aquarius exhibits structural adaptations mediating its recruitment to spliceosomes.
The RNA helicase Aquarius exhibits structural adaptations mediating its recruitment to spliceosomes.
Nature structural & molecular biology (2015-01-20)
Inessa De, Sergey Bessonov, Romina Hofele, Karine dos Santos, Cindy L Will, Henning Urlaub, Reinhard Lührmann, Vladimir Pena
ABSTRACT
Aquarius is a multifunctional putative RNA helicase that binds precursor-mRNA introns at a defined position. Here we report the crystal structure of human Aquarius, revealing a central RNA helicase core and several unique accessory domains, including an ARM-repeat domain. We show that Aquarius is integrated into spliceosomes as part of a pentameric intron-binding complex (IBC) that, together with the ARM domain, cross-links to U2 snRNP proteins within activated spliceosomes; this suggests that the latter aid in positioning Aquarius on the intron. Aquarius's ARM domain is essential for IBC formation, thus indicating that it has a key protein-protein-scaffolding role. Finally, we provide evidence that Aquarius is required for efficient precursor-mRNA splicing in vitro. Our findings highlight the remarkable structural adaptations of a helicase to achieve position-specific recruitment to a ribonucleoprotein complex and reveal a new building block of the human spliceosome.
Sigma-Aldrich
Anti-ISY1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution