Merck
  • 2-(3-Aryl-2-propenoyl)-3-methylquinoxaline-1,4-dioxides: a novel cluster of tumor-specific cytotoxins which reverse multidrug resistance.
2-(3-Aryl-2-propenoyl)-3-methylquinoxaline-1,4-dioxides: a novel cluster of tumor-specific cytotoxins which reverse multidrug resistance.
Bioorganic & medicinal chemistry (2009-05-12)
Umashankar Das, Hari N Pati, Atulya K Panda, Erik De Clercq, Jan Balzarini, Joseph Molnár, Zoltán Baráth, Imre Ocsovszki, Masami Kawase, Li Zhou, Hiroshi Sakagami, Jonathan R Dimmock
ABSTRACT
A series of 2-(3-aryl-2-propenoyl)-3-methylquinoxaline-1,4-dioxides 3a-l were prepared by condensation of various aryl aldehydes with 2-acetyl-3-methylquinoxaline-1,4-dioxide 2. These compounds inhibit the growth of human Molt 4/C8 and CEM T-lymphocytes and the IC(50) values are mainly in the 5-30 microM range. The quinoxaline 1,4-dioxide 3j inhibited the growth of 58 human tumor cell lines, particularly leukemic and breast cancer neoplasms. All of the compounds 3a-l reversed the multidrug resistance (MDR) properties of murine L-5178Y leukemic cells which were transfected with the human MDR1 gene. The MDR-reversing effect may be due to the conjugated pi-electron system forming a weak electron charge transfer complex with the P-glycoprotein-mediated efflux pump. The compounds in series 2 and 3 were assessed against HL-60, HSC-2, HSC-3 and HSC-4 malignant cells as well as HGF, HPC and HPLF normal cell lines which revealed that the majority of the compounds displayed a greater toxicity to neoplastic than normal cells. Various ways in which the project may be expanded are presented.
Sigma-Aldrich
2-Methylquinoxaline, 97%
Sigma-Aldrich
2-Methylquinoxaline, ≥97%
Sigma-Aldrich
Melphalan, powder
Supelco
Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
Supelco
Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Discovery® C8 Supelguard Guard Cartridge Kit, 5 μm particle size, L × I.D. 2 cm × 4 mm, contains nuts, ferrules, guard holder