Merck
  • Methane Ameliorates Lipopolysaccharide-Induced Acute Orchitis by Anti-inflammatory, Antioxidative, and Antiapoptotic Effects via Regulation of the PK2/PKR1 Pathway.
Methane Ameliorates Lipopolysaccharide-Induced Acute Orchitis by Anti-inflammatory, Antioxidative, and Antiapoptotic Effects via Regulation of the PK2/PKR1 Pathway.
Oxidative medicine and cellular longevity (2020-09-15)
Chao Huang, Wenbo Zhang, Aijun Sun, Xi Zhang, Jinping Guo, Ruijuan Ji, Liang Qiao, Xuejun Sun, Dongbao Zhao
ABSTRACT
The present study is aimed at investigating the anti-inflammatory, antioxidative, and antiapoptotic effects of methane on lipopolysaccharide- (LPS-) induced acute orchitis and its potential mechanisms. Adult male rats were intraperitoneally (i.p.) injected with methane-rich saline (MS, 20 mL/kg) following LPS (5 mg/kg, i.p.). The survival rate was assessed every 12 h until 72 h after LPS induction, and surviving rats were sacrificed for further detection. The wet/dry (W/D) ratio was determined, and testicular damage was histologically assessed. Inflammatory cytokines in the testes and serum, including interleukin-1β (IL-1β), IL-6, IL-10, and tumor necrosis factor-α (TNF-α), were measured using ELISA and RT-qPCR. Oxidative stress was evaluated by the level of superoxide dismutase (SOD) and malondialdehyde (MDA). Testicular apoptosis was detected via TUNEL staining. The expression of prokineticin 2 (PK2)/prokineticin receptor 1 (PKR1) was also analyzed using RT-qPCR, western blotting, and immunohistochemistry. It was found that methane significantly prolonged rat survival, decreased the W/D ratio, alleviated LPS-induced histological changes, and reduced apoptotic cells in the testes. Additionally, methane suppressed and promoted the production of pro- and anti-inflammatory cytokines, respectively. Furthermore, methane significantly increased SOD levels, decreased MDA levels, and decreased testicular expression of PK2 and PKR1. Therefore, methane exerts therapeutic effects on acute orchitis and might be a new and convenient strategy for the treatment of inflammation-related testicular diseases.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction