Merck
  • The association of pili with the emergence and replacement of the major antibiotic resistant pneumococcal clones.
The association of pili with the emergence and replacement of the major antibiotic resistant pneumococcal clones.
Journal of microbiology, immunology, and infection = Wei mian yu gan ran za zhi (2019-04-24)
Helena Zemlickova, Vladislav Jakubu, Marta Fridrichova, Lucia Malisova, Martin Musilek, Milan Trojanek
ABSTRACT
The aim of this study was to evaluate the presence of pilus islet 1 (PI-1) and to determine its clade type in pneumococcal isolates with reduced susceptibility to penicillin (penicillin non-susceptible pneumococci - PNSP) and/or resistant to macrolides isolated prior to and after the introduction of pneumococcal conjugate vaccines (PCVs) in the Czech Republic. Clinical isolates of serotypes 9V (n = 68) and 19A (n = 89) were examined. Isolates were characterised by multilocus sequence typing (MLST). The presence of PI-1 was determined by screening for the sortase B, C, and D genes located within PI-1. In the presence of PI-1 pilus, clade types were classified by PCR. In the pre-PCV period (2000-2007), the prevalence of PNSP was 3.9% and 2.7% of isolates were resistant to erythromycin. During 2012-2015 (post-PCV period), the rates of PNSP remained stable (3.6%), but resistance to erythromycin increased to 8.3%. While in 2000-2007, resistance to antibiotics was associated mainly with serotype 9V, in 2012-2015, it was replaced by serotype 19A. PI-1 positive isolates were seen in both serotypes. All isolates (68) of serotype 9V belonged to the Spain9V-3 (CC156) clone and carried PI-1 of clade type I while 96.5% (56/58) of isolates of 19A serotype belonged to the Netherlands15B-37 (CC199) clone and carried PI-1 of clade type II. Both major antibiotic resistant clones carried PI-1, although they differ in the clade type. Thus the role of PI-1 should be evaluated in further studies and potentially considered in the spread of antibiotic resistant clones.
Sigma-Aldrich
GenElute Bacterial Genomic DNA Kits, sufficient for 350 purifications
Sigma-Aldrich
Mutanolysin from Streptomyces globisporus ATCC 21553, lyophilized powder, ≥4000 units/mg protein (biuret), Chromatographically purified