Merck
  • A divergent mode of activation of a nitrosyl iron complex with unusual antiangiogenic activity.
A divergent mode of activation of a nitrosyl iron complex with unusual antiangiogenic activity.
Journal of inorganic biochemistry (2020-07-04)
Edinilton Muniz Carvalho, Lisa A Ridnour, Florêncio Sousa Gouveia Júnior, Pedro Henrique Bezerra Cabral, Nilberto Robson Falcão do Nascimento, David A Wink, Douglas W Franco, Mayara Jane Campos de Medeiros, Daniel de Lima Pontes, Elisane Longhinotti, Tércio de Freitas Paulo, Vania Bernardes-Génisson, Remi Chauvin, Eduardo Henrique Silva Sousa, Luiz Gonzaga de França Lopes
ABSTRACT
Nitric oxide (NO) and nitroxyl (HNO) have gained broad attention due to their roles in several physiological and pathophysiological processes. Remarkably, these sibling species can exhibit opposing effects including the promotion of angiogenic activity by NO compared to HNO, which blocks neovascularization. While many NO donors have been developed over the years, interest in HNO has led to the recent emergence of new donors. However, in both cases there is an expressive lack of iron-based compounds. Herein, we explored the novel chemical reactivity and stability of the trans-[Fe(cyclam)(NO)Cl]Cl2 (cyclam = 1,4,8,11-tetraazacyclotetradecane) complex. Interestingly, the half-life (t1/2) for NO release was 1.8 min upon light irradiation, vs 5.4 h upon thermal activation at 37 °C. Importantly, spectroscopic evidence supported the generation of HNO rather than NO induced by glutathione. Moreover, we observed significant inhibition of NO donor- or hypoxia-induced HIF-1α (hypoxia-inducible factor 1α) accumulation in breast cancer cells, as well as reduced vascular tube formation by endothelial cells pretreated with the trans-[Fe(cyclam)(NO)Cl]Cl2 complex. Together, these studies provide the first example of an iron-nitrosyl complex with anti-angiogenic activity as well as the potential dual activity of this compound as a NO/HNO releasing agent, which warrants further pharmacological investigation.
Sigma-Aldrich
1,4,8,11-Tetraazacyclotetradecane, 98% (GC)