Merck
  • Structurally diverse halosesquiterpenoids from the red alga Laurencia composita Yamada.
Structurally diverse halosesquiterpenoids from the red alga Laurencia composita Yamada.
Fitoterapia (2020-09-01)
Zhi-Biao Hu, Xiao-Qing Yu, Bin Wang, Ai-Hong Liu, Tian-Sheng Zhao, Yue-Wei Guo, Hui-Lian Huang, Shui-Chun Mao
ABSTRACT
A phytochemical investigation on the MeOH extract of the red alga Laurencia composita Yamada led to the discovery of six new highly halogenated sesquiterpenoids, including two bisabolane-type sesquiterpenoids (1 and 2), one nerolidol derivative (7), and three chamigrane-type sesquiterpenoids (9, 10, and 18), together with 13 known sesquiterpenoids. Their structures, including relative configuration, were elucidated by extensive spectroscopic analysis, and by comparison with data for related known compounds. The absolute configuration at C-10 of laurecomposin A (1) was determined by the modified Mosher's method. Halonerolidol (7) is the first naturally occurring halogenated nerolidol derivative, while compositacin L (9) represents the third example of chamigranes having a C-10 carbonyl group. Antifungal, antibacterial, and receptor tyrosine kinase inhibitory activities of these isolates were evaluated. The results showed that compounds 1-3 and 5 exhibited significant antifungal activity against Microsporum gypseum (Cmccfmza) with MIC values of 4, 8, 8, and 4 μg/mL, respectively. Additionally, compounds 1-3 and 5 also displayed promising antibacterial activity against Gram-positive bacteria Staphylococcus aureus Newman strain with MIC values ranging from 10.9 to 26.8 μg/mL.
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)