Merck
  • Host-derived circular RNAs display proviral activities in Hepatitis C virus-infected cells.
Host-derived circular RNAs display proviral activities in Hepatitis C virus-infected cells.
PLoS pathogens (2020-08-09)
Tzu-Chun Chen, Marc Tallo-Parra, Qian M Cao, Sebastian Kadener, René Böttcher, Gemma Pérez-Vilaró, Pakpoom Boonchuen, Kunlaya Somboonwiwat, Juana Díez, Peter Sarnow
ABSTRACT
Viruses subvert macromolecular pathways in infected host cells to aid in viral gene amplification or to counteract innate immune responses. Roles for host-encoded, noncoding RNAs, including microRNAs, have been found to provide pro- and anti-viral functions. Recently, circular RNAs (circRNAs), that are generated by a nuclear back-splicing mechanism of pre-mRNAs, have been implicated to have roles in DNA virus-infected cells. This study examines the circular RNA landscape in uninfected and hepatitis C virus (HCV)-infected liver cells. Results showed that the abundances of distinct classes of circRNAs were up-regulated or down-regulated in infected cells. Identified circRNAs displayed pro-viral effects. One particular up-regulated circRNA, circPSD3, displayed a very pronounced effect on viral RNA abundances in both hepatitis C virus- and Dengue virus-infected cells. Though circPSD3 has been shown to bind factor eIF4A3 that modulates the cellular nonsense-mediated decay (NMD) pathway, circPSD3 regulates RNA amplification in a pro-viral manner at a post-translational step, while eIF4A3 exhibits the anti-viral property of the NMD pathway. Findings from the global analyses of the circular RNA landscape argue that pro-, and likely, anti-viral functions are executed by circRNAs that modulate viral gene expression as well as host pathways. Because of their long half-lives, circRNAs likely play hitherto unknown, important roles in viral pathogenesis.
Sigma-Aldrich
Hexarelin, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human PSD3