Merck
  • Lactate predicts the 28-day survival rate in patients with septic shock treated with the combination of PMX-DHP and rTM.
Lactate predicts the 28-day survival rate in patients with septic shock treated with the combination of PMX-DHP and rTM.
Therapeutic apheresis and dialysis : official peer-reviewed journal of the International Society for Apheresis, the Japanese Society for Apheresis, the Japanese Society for Dialysis Therapy (2020-06-12)
Masafumi Yamato, Yuta Asahina, Shintaro Koizumi, Takatomo Shigeki, Ayako Yajima, Yoshiki Kimura, Hirotsugu Iwatani
ABSTRACT
We have previously reported that combination therapy with polymyxin-B direct hemoperfusion (PMX-DHP) and recombinant thrombomodulin (rTM) is effective in patients with septic shock accompanied by disseminated intravascular coagulation (DIC). Two previous studies reporting the favorable effect of early initiation of PMX-DHP for septic shock did not focus on the combination therapy of PMX-DHP and rTM. This retrospective study included 47 consecutive patients who underwent the combination therapy of PMX-DHP and rTM for septic shock with DIC from August 2011 to August 2016. Main exposure was early or late initiation of PMX-DHP. PMX-DHP initiated within 12 hours after catecholamine administration was designated as early group (N = 25) and later than 12 hours as late group (N = 22). Main outcome was 28-day survival rate. The patient characteristics were age median 73 (IQR 68-78) years, 26 men (55%), APACHE II score 32.7 ± 7.7 and lactate 26.0 (18.0-41.0) mg/dL. The 28-day survival rate after PMX-DHP initiation was 76.6% and was not significantly different in the two groups. In the early group, APACHE II score was lower (P = .02), and lactate was higher (P = .005) than in the late group. Lactate was the only predictor of 28-day mortality [odds ratio (95%CI) per 1 mg/dL, 1.08 (1.03-1.19); P = .037] in multivariate logistic regression analysis adjusted with age, sex, APACHE II score, lactate and timing of PMX-DHP initiation. Late PMX-DHP initiation did not lead to statistically worse 28-day survival rate in this combination therapy. The combination therapy of PMX-DHP and rTM may improve the therapeutic effect of PMX-DHP and modify the effect of early PMX-DHP on the prognosis. Lactate may be an appropriate indicator rather than time after catecholamine administration if we discuss when to start PMX-DHP in this combination therapy.
Sigma-Aldrich
Carboxyamidotriazole, ≥98% (HPLC)