Merck
  • Effects of Synbiotic Supplementation on Chronic Inflammation and the Gut Microbiota in Obese Patients with Type 2 Diabetes Mellitus: A Randomized Controlled Study.
Effects of Synbiotic Supplementation on Chronic Inflammation and the Gut Microbiota in Obese Patients with Type 2 Diabetes Mellitus: A Randomized Controlled Study.
Nutrients (2021-02-12)
Akio Kanazawa, Masanori Aida, Yasuto Yoshida, Hideyoshi Kaga, Takehiro Katahira, Luka Suzuki, Shoko Tamaki, Junko Sato, Hiromasa Goto, Kosuke Azuma, Tomoaki Shimizu, Takuya Takahashi, Yuichiro Yamashiro, Hirotaka Watada
ABSTRACT
The aim of this study was to investigate the effects of 24-week synbiotic supplementation on chronic inflammation and the gut microbiota in obese patients with type 2 diabetes. We randomized 88 obese patients with type 2 diabetes to one of two groups for 24 weeks: control or synbiotic (Lacticaseibacillus paracasei strain Shirota (previously Lactobacillus casei strain Shirota) and Bifidobacterium breve strain Yakult, and galactooligosaccharides). The primary endpoint was the change in interleukin-6 from baseline to 24 weeks. Secondary endpoints were evaluation of the gut microbiota in feces and blood, fecal organic acids, high-sensitivity C-reactive protein, lipopolysaccharide-binding protein, and glycemic control. Synbiotic administration for 24 weeks did not significantly affect changes in interleukin-6 from baseline to 24 weeks (0.35 ± 1.99 vs. -0.24 ± 1.75 pg/mL, respectively). Relative to baseline, however, at 24 weeks after synbiotic administration there were positive changes in the counts of Bifidobacterium and total lactobacilli, the relative abundances of Bifidobacterium species such as Bifidobacterium adolescentis and Bifidobacterium pseudocatenulatum, and the concentrations of acetic and butyric acids in feces. No significant changes in inflammatory markers were found in the synbiotic group compared to the control group. However, synbiotic administration at least partially improved the gut environment in obese patients with type 2 diabetes.
Sigma-Aldrich
Human LBP ELISA Kit, for cell culture supernatants, plasma, and serum samples