Merck
  • Gut flora-targeted photobiomodulation therapy improves senile dementia in an Aß-induced Alzheimer's disease animal model.
Gut flora-targeted photobiomodulation therapy improves senile dementia in an Aß-induced Alzheimer's disease animal model.
Journal of photochemistry and photobiology. B, Biology (2021-02-21)
Qianqian Chen, Jinpeng Wu, Xiaoxi Dong, Huijuan Yin, Xiafei Shi, Siying Su, Bochen Che, Yingxin Li, Jichun Yang
ABSTRACT
Emerging evidence suggests that the gut microbiota plays an important role in the pathological progression of Alzheimer's disease (AD). Photobiomodulation (PBM) therapy is believed to have a positive regulatory effect on the imbalance of certain body functions, including inflammation, immunity, wound healing, nerve repair, and pain. Previous studies have found that the intestinal flora of patients with AD is in an unbalanced state. Therefore, we have proposed the use of gut flora-targeted PBM (gf-targeted PBM) as a method to improve AD in an Aß-induced AD mouse model. PBM was performed on the abdomen of the mice at the wavelengths of 630 nm, 730 nm, and 850 nm at 100 J/cm2 for 8 weeks. Morris water maze test, immunofluorescence and proteomic of hippocampus, and intestinal flora detection of fecal were used to evaluate the treatment effects of gf-targeted PBM on AD rats. PBM at all three wavelengths (especially 630 nm and 730 nm) significantly improved learning retention as measured by the Morris water maze. In addition, we found reduced amyloidosis and tau phosphorylation in the hippocampus by immunofluorescence in AD mice. By using a quantitative proteomic analysis of the hippocampus, we found that gf-targeted PBM significantly altered the expression levels of 509 proteins (the same differentially expressed proteins in all three wavelengths of PBM), which involved the pathways of hormone synthesis, phagocytosis, and metabolism. The 16 s rRNA gene sequencing of fecal contents showed that PBM significantly altered the diversity and abundance of intestinal flora. Specifically, PBM treatment reversed the typical increase of Helicobacter and uncultured Bacteroidales and the decrease of Rikenella seen in AD mice. Our data indicate that gf-targeted PBM regulates the diversity of intestinal flora, which may improve damage caused by AD. Gf-targeted PBM has the potential to be a noninvasive microflora regulation method for AD patients.
Sigma-Aldrich
Mouse IFN-γ ELISA Kit, for serum, plasma, cell culture supernatant
Pricing and availability is not currently available.
Sigma-Aldrich
Mouse IL-6 ELISA Kit, for serum, plasma and cell culture supernatant
Pricing and availability is not currently available.