Merck
Genome Sequence for Candida albicans Clinical Oral Isolate 529L.
Microbiology resource announcements (2019-06-22)
Christina A Cuomo, Saranna Fanning, Sharvari Gujja, Qiandong Zeng, Julian R Naglik, Scott G Filler, Aaron P Mitchell
ABSTRACT
The diploid heterozygous yeast Candida albicans is the most common cause of fungal infection. Here, we report the genome sequence assembly of the clinical oral isolate 529L. As this isolate grows as a commensal, this genome will serve as a reference for experimental and genetic studies of mucosal colonization.
Sigma-Aldrich
Lyticase from Arthrobacter luteus, lyophilized powder, ≥2,000 units/mg protein, Protein ≥20 % by biuret