Merck
  • miR-17-5p drives G2/M-phase accumulation by directly targeting CCNG2 and is related to recurrence of head and neck squamous cell carcinoma.
miR-17-5p drives G2/M-phase accumulation by directly targeting CCNG2 and is related to recurrence of head and neck squamous cell carcinoma.
BMC cancer (2021-10-03)
Qiang Huang, Yu-Jie Shen, Chi-Yao Hsueh, Yang Guo, Yi-Fan Zhang, Jiao-Yu Li, Liang Zhou
ABSTRACT
The human miR-17-92 polycistron is the first reported and most well-studied onco-miRNA with a cluster of seven miRNAs. miR-17-5p, a member of the miR-17-92 family, plays an important role in tumor cell proliferation, apoptosis, migration and invasion. However, few studies have shown the role of miR-17-5p in the cell cycle of head and neck squamous cell carcinoma (HNSCC). RT-qPCR was used to detect miR-17-5p expression levels in 64 HNSCC tissues and 5 cell lines. The relationship between the expression of miR-17-5p in the tissues and the clinical characteristics of the patients was analyzed. HNSCC cells were transfected with an miR-17-5p mimic or inhibitor to evaluate cell cycle distribution by flow cytometry. Cell cycle distribution of cells transfected with target gene was evaluated using flow cytometry. Dual-luciferase reporter assay was used to detect the regulatory effect of miR-17-5p on target gene expression. In the present study, we found that miR-17-5p expression in HNSCC tissues and cell lines was remarkably increased, and miR-17-5p is related to recurrence in HNSCC patients. Silencing miR-17-5p blocked HNSCC cells in G2/M phase, whereas its overexpression propelled cell cycle progression. More importantly, we verified that miR-17-5p negatively regulated CCNG2 mRNA and protein expression by directly targeting its 3'UTR. These findings suggest that miR-17-5p might act as a tumor promoter and prognostic factor for recurrence in HNSCC patients.