Merck
  • Adipose stem cell sheets improved cardiac function in the rat myocardial infarction, but did not alter cardiac contractile responses to β-adrenergic stimulation.
Adipose stem cell sheets improved cardiac function in the rat myocardial infarction, but did not alter cardiac contractile responses to β-adrenergic stimulation.
Biomedical research (Tokyo, Japan) (2015-03-10)
Yuki Otsuki, Yoshinobu Nakamura, Shingo Harada, Yasutaka Yamamoto, Kazuhide Ogino, Kumi Morikawa, Haruaki Ninomiya, Shigeru Miyagawa, Yoshiki Sawa, Ichiro Hisatome, Motonobu Nishimura
ABSTRACT
Adipose stem cells (ASCs) are a source of regenerative cells available for autologous transplantation to hearts. We compared protective actions of ASC sheets on rat myocardial infarction (MI) in comparison with those of skeletal myoblast cell sheets. Their effects on infarcted hearts were evaluated by biological, histochemical as well as physiological analyses. ASC sheets secreted higher concentrations of angiogenic factors (HGF, VEGF, and bFGF; P < 0.05) under normoxic and hypoxic conditions than those of myoblast cell sheets, associated with reduction of cell apoptosis (P < 0.05). Like myoblast cell sheets, ASC sheets improved cardiac function (P < 0.05) and decreased the plasma level of ANP (P < 0.05) in MI hearts. ASC sheets restored cardiac remodeling characterized by fibrosis, cardiac hypertrophy and impaired angiogenesis (P < 0.05), which was associated with increases in angiogenic factors (P < 0.05). In isolated perfused rat hearts, ASC sheets improved both systolic and diastolic functions, which was comparable to cardiac functions of myoblast cell sheets, while both cell sheets failed to restore cardiac contractile response to either isoproterenol, pimobendan or dibutyryl cAMP. These results indicated that ASC sheets improved cardiac function and remodeling of MI hearts mediated by their paracrine action and this improvement was comparable to those by myoblast cell sheets.
Sigma-Aldrich
Anti-Tropomyosin (Sarcomeric) antibody, Mouse monoclonal, clone CH1, purified from hybridoma cell culture