Merck
  • Genome Sequence of the Oleaginous Yeast Rhodotorula glutinis ATCC 204091.
Genome Sequence of the Oleaginous Yeast Rhodotorula glutinis ATCC 204091.
Genome announcements (2014-02-15)
Debarati Paul, Zenaida Magbanua, Mark Arick, Todd French, Susan M Bridges, Shane C Burgess, Mark L Lawrence
ABSTRACT
Rhodotorula glutinis ATCC 204091 is an oleaginous oxidative red yeast that can accumulate lipids to >50% of its biomass when grown with appropriate carbon and nitrogen ratios. It produces a red pigment consisting of useful antioxidants, such as carotenoids, torulene, and torularhodin, when cultivated under carbon-deficient conditions.
Sigma-Aldrich
Lyticase from Arthrobacter luteus, lyophilized powder, ≥2,000 units/mg protein, Protein ≥20 % by biuret