Merck
  • Prostaglandin E2-EP4 Axis Promotes Lipolysis and Fibrosis in Adipose Tissue Leading to Ectopic Fat Deposition and Insulin Resistance.
Prostaglandin E2-EP4 Axis Promotes Lipolysis and Fibrosis in Adipose Tissue Leading to Ectopic Fat Deposition and Insulin Resistance.
Cell reports (2020-10-15)
Tomoaki Inazumi, Kiyotaka Yamada, Naritoshi Shirata, Hiroyasu Sato, Yoshitaka Taketomi, Kazunori Morita, Hirofumi Hohjoh, Soken Tsuchiya, Kentaro Oniki, Takehisa Watanabe, Yutaka Sasaki, Yuichi Oike, Yasuhiro Ogata, Junji Saruwatari, Makoto Murakami, Yukihiko Sugimoto
ABSTRACT
Lipolysis, the breakdown of triglyceride storage in white adipose tissue, supplies fatty acids to other tissues as a fuel under fasting conditions. In morbid obesity, fibrosis limits adipocyte expandability, resulting in enforced lipolysis, ectopic fat distribution, and ultimately insulin resistance. Although basal levels of lipolysis persist even after feeding, the regulatory mechanisms of basal lipolysis remain unclear. Here, we show the important role of adipocyte prostaglandin (PG) E2-EP4 receptor signaling in controlling basal lipolysis, fat distribution, and collagen deposition during feeding-fasting cycles. The PGE2-synthesis pathway in adipocytes, which is coupled with lipolysis, is activated by insulin during feeding. By regulating the lipolytic key players, the PGE2-EP4 pathway sustains basal lipolysis as a negative feedback loop of insulin action, and perturbation of this process leads to "metabolically healthy obesity." The potential role of the human EP4 receptor in lipid regulation was also suggested through genotype-phenotype association analyses.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, L-glutamine, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
6-Bnz-cAMP sodium salt, ≥98% (HPLC)
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Glycerol Assay Kit, sufficient for 200 colorimetric or fluorometric tests
Millipore
SB 202190, SB 202190, CAS 152121-30-7, is a potent, reversible, competitive inhibitor of p38. Inhibits p38 phosphorylation of myelin basic protein. Blocks the activity of p38β (Ki = 16 nM; IC50 = 350 nM).
Sigma-Aldrich
(−)-Isoproterenol hydrochloride
Sigma-Aldrich
Dexamethasone-Water Soluble, suitable for cell culture, BioReagent