Merck
  • Sympathetic signaling facilitates progression of neuroendocrine prostate cancer.
Sympathetic signaling facilitates progression of neuroendocrine prostate cancer.
Cell death discovery (2021-11-24)
Shubham Dwivedi, Maricris Bautista, Sanskriti Shrestha, Hussain Elhasasna, Tanaya Chaphekar, Frederick S Vizeacoumar, Anand Krishnan
ABSTRACT
The progression of prostate cancer (PC) into neuroendocrine prostate cancer (NEPC) is a major challenge in treating PC. In NEPC, the PC cells undergo neuroendocrine differentiation (NED); however, the exact molecular mechanism that triggers NED is unknown. Peripheral nerves are recently shown to promote PC. However, their contribution to NEPC was not studied well. In this study, we explored whether sympathetic neurosignaling contributes to NED. We found that human prostate tumors from patients that later developed metastases and castration-resistant prostate cancer (CRPC), a stage preceding to NEPC, have high sympathetic innervations. Our work revealed that high concentrations of the sympathetic neurotransmitter norepinephrine (NE) induces NED-like changes in PC cells in vitro, evident by their characteristic cellular and molecular changes. The NE-mediated NED was effectively inhibited by the Adrβ2 blocker propranolol. Strikingly, propranolol along with castration also significantly inhibited the development and progression of NEPC in vivo in an orthotopic NEPC model. Altogether, our results indicate that the NE-Adrβ2 axis is a potential therapeutic intervention point for NEPC.
Sigma-Aldrich
Anti-Beta III Tubulin Antibody, Chemicon®, from chicken
Sigma-Aldrich
L-(−)-Norepinephrine (+)-bitartrate salt monohydrate, ≥99%, solid
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit