Merck
  • Steroid Receptor Coregulators Can Modulate the Action of Progesterone Receptor during the Estrous Cycle in Cow Endometrium.
Steroid Receptor Coregulators Can Modulate the Action of Progesterone Receptor during the Estrous Cycle in Cow Endometrium.
Animals : an open access journal from MDPI (2021-11-28)
Robert Rekawiecki, Karolina Dobrzyn, Magdalena K Kowalik
ABSTRACT
Nuclear receptor coregulators include coactivators and corepressors which associate with the progesterone receptor (PGR) during its activation. Fluctuations in the transcription levels of their respective genes and subsequent protein production as well as in related activities for histone acetyltransferase (HAT) and histone deacetylase (HDAC) can affect PGR function and thus change the action of progesterone (P4) in bovine endometrium during the estrous cycle. Endometrial tissue on days 2-5, 6-10, 11-16, and 17-20 of the estrous cycle was used for determination of the mRNA expression levels of coactivators P300, CREB, and SRC-1 along with corepressor NCOR-2 using Real-Time PCR, with protein levels by Western blot. Coregulators cellular localizations were assessed by immunohistochemistry whereas the activities of HAT and HDAC by using EIA. The highest levels of mRNA and proteins for all of the investigated coregulators, as well as the highest levels of activity for HAT and HDAC, were detected over days 2-16 of the estrous cycle. All of the tested coregulatory proteins were localized in the nuclei of endometrial cells. This research indicates the important role of coregulators of the PGR receptor in regulating P4 activity in endometrial cells, especially during the pre-implantation period.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-NCOA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody