Merck
  • Regulation of adipose tissue lipolysis by ghrelin is impaired with high-fat diet feeding and is not restored with exercise.
Regulation of adipose tissue lipolysis by ghrelin is impaired with high-fat diet feeding and is not restored with exercise.
Adipocyte (2021-07-06)
Barbora Hucik, Andrew J Lovell, Evan M Hoecht, Daniel T Cervone, David M Mutch, David J Dyck
ABSTRACT
Ghrelin is released from the stomach as an anticipatory signal prior to a meal and decreases immediately after. Previous research has shown that both acylated (AG) and unacylated (UnAG) ghrelin blunt adrenoreceptor-stimulated lipolysis in rat white adipose tissue (WAT) ex vivo. We investigated whether acute or chronic consumption of a high fat diet (HFD) impaired the ability of ghrelin to regulate adipose tissue lipolysis, and if this impairment could be restored with exercise. After 5 days (5d) of a HFD, or 6 weeks (6 w) of a HFD (60% kcal from fat) with or without exercise training, inguinal and retroperitoneal WAT was collected from anesthetized rats for adipose tissue organ culture. Samples were treated with 1 μM CL 316,243 (CL; lipolytic control), 1 μM CL+150 ng/ml AG or 1 μM CL+150 ng/ml UnAG. Incubation media and tissue were collected after 2 hours. Colorometric assays were used to determine glycerol and free fatty acid (FFA) concentrations in media. Western blots were used to quantify the protein content of lipolytic enzymes and ghrelin receptors in both depots. CL stimulated lipolysis was evidenced by increases in glycerol (p < 0.0001) and FFA (p < 0.0001) concentrations in media compared to control. AG decreased CL-stimulated glycerol release in inguinal WAT from 5d LFD rats (p = 0.0097). Neither AG nor UnAG blunted lipolysis in adipose tissue from 5d or 6 w HFD-fed rats, and exercise did not restore ghrelin's anti-lipolytic ability in 6 w HFD-fed rats. Overall, this study demonstrates that HFD consumption impairs ghrelin's ability to regulate adipose tissue lipolysis.
Sigma-Aldrich
Glycerol Standard Solution, 2.5 mg/ml equivalent triolein concentration
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol