Merck
Roles of matrix metalloproteinase-7 (MMP-7) in cancer.
Clinical biochemistry (2021-03-14)
Hai-Yang Liao, Chao-Ming Da, Bei Liao, Hai-Hong Zhang
ABSTRACT
Matrix metalloproteinase-7 (MMP-7) is a small proteolytic enzyme that secretes zinc and calcium endopeptidases. It can degrade a variety of extracellular matrix substrates and other substrates and plays important regulatory roles in many human pathophysiological processes. Since its discovery, MMP-7 has been recognized as a regulatory protein in wound healing, bone growth, and remodeling. Later, MMP-7 was reported to regulate the occurrence and development of cancers and mediate the proliferation, differentiation, metastasis, and invasion of several types of cancer cells via various mechanisms. Thus, matrix metalloproteinase-7 may be a promising tumor biomarker and therapeutic target. The expression of MMP-7 correlates with the clinical characteristics of cancer patients, and its expression profile is a new diagnostic and prognostic biomarker for a variety of human diseases. Hence, manipulating the expression or function of MMP-7 may be a potential treatment strategy for different diseases including cancers. This review summarizes the role played by MMP-7 in carcinogenesis of several human cancers, underlying mechanisms, and its clinical significance of the occurrence and development of cancers.
Sigma-Aldrich
Human MMP-7 ELISA Kit, on for cell culture supernatant
Pricing and availability is not currently available.