Merck
  • Hierarchical macro-mesoporous structures in the system TiO2-Al2O3, obtained by hydrothermal synthesis using Tween-20 as a directing agent
Hierarchical macro-mesoporous structures in the system TiO2-Al2O3, obtained by hydrothermal synthesis using Tween-20 as a directing agent
Materials Characterization (2009)
Garcia-Benjume ML, et al.