Merck
  • Design, synthesis, in silico studies and in vitro evaluation of isatin-pyridine oximes hybrids as novel acetylcholinesterase reactivators.
Design, synthesis, in silico studies and in vitro evaluation of isatin-pyridine oximes hybrids as novel acetylcholinesterase reactivators.
Journal of enzyme inhibition and medicinal chemistry (2021-06-22)
Daniel A S Kitagawa, Rafael B Rodrigues, Thiago N Silva, Wellington V Dos Santos, Vinicius C V da Rocha, Joyce S F D de Almeida, Leandro B Bernardo, Taynara Carvalho-Silva, Cintia N Ferreira, Angelo A T da Silva, Alessandro B C Simas, Eugenie Nepovimova, Kamil Kuča, Tanos C C França, Samir F de A Cavalcante
ABSTRACT
Organophosphorus poisoning caused by some pesticides and nerve agents is a life-threating condition that must be swiftly addressed to avoid casualties. Despite the availability of medical countermeasures, the clinically available compounds lack a broad spectrum, are not effective towards all organophosphorus toxins, and have poor pharmacokinetics properties to allow them crossing the blood-brain barrier, hampering cholinesterase reactivation at the central nervous system. In this work, we designed and synthesised novel isatin derivatives, linked to a pyridinium 4-oxime moiety by an alkyl chain with improved calculated properties, and tested their reactivation potency against paraoxon- and NEMP-inhibited acetylcholinesterase in comparison to the standard antidote pralidoxime. Our results showed that these compounds displayed comparable in vitro reactivation also pointed by the in silico studies, suggesting that they are promising compounds to tackle organophosphorus poisoning.
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type V-S, lyophilized powder, ≥1,000 units/mg protein
Pricing and availability is not currently available.