Merck
  • Salinomycin triggers prostate cancer cell apoptosis by inducing oxidative and endoplasmic reticulum stress via suppressing Nrf2 signaling.
Salinomycin triggers prostate cancer cell apoptosis by inducing oxidative and endoplasmic reticulum stress via suppressing Nrf2 signaling.
Experimental and therapeutic medicine (2021-07-27)
Jianyong Yu, Yang Yang, Shan Li, Peng Meng
ABSTRACT
Salinomycin is a polyether antiprotozoal antibiotic that is widely used as an animal food additive. Some antifungal, antiparasitic, antiviral and anti-inflammatory activities have been reported for salinomycin. Recently, the anti-cancer effect of salinomycin has been demonstrated in breast cancer; however, the underlying mechanism remains unknown. The present study aimed to investigate the functional roles of salinomycin in the progression of prostate cancer cells using the DU145 and PC-3 cell lines. Western blotting and reverse transcription-quantitative polymerase chain reaction were performed to detect the expression of oxidative stress and endoplasmic reticulum stress-related molecules, and flow cytometry was performed to detect the apoptosis rate of DU145 and PC-3 cells after salinomycin treatment. The results demonstrated that salinomycin inhibited the viability and induced the apoptosis of PC-3 and DU145 cells in a dose-dependent manner. Furthermore, salinomycin increased the production of reactive oxygen species (ROS) and 8-hydroxy-2'-deoxyguanosine (8-OH-dG) and the lipid peroxidation. In addition, salinomycin induced the activation of unfolded protein response and endoplasmic reticulum stress in DU145 and PC-3 cells, as indicated by the elevated expression of binding immunoglobulin protein, activating transcription factor 4, phosphorylated eukaryotic initiation factor 2α, phosphorylated protein kinase RNA-like endoplasmic reticulum kinase and C/EBP homologous protein. In addition, salinomycin significantly downregulated the expression of nuclear factor erythroid 2-related factor 2 (Nrf2), heme oxygenase-1, NAD(P)H quinone dehydrogenase 1 and glutamate-cysteine ligase catalytic subunit and decreased the activity of the antioxidant enzymes superoxide dismutase, catalase and glutathione peroxidase in PC-3 and DU145 cells. Furthermore, the Nrf2 activator, tert-butylhydroquinone, significantly reversed the therapeutic effects of salinomycin by stimulating the Nrf2 pathway and increasing the activity of antioxidant enzymes. Taken together, these findings demonstrated that salinomycin may trigger apoptosis by inducing oxidative and ER stress in prostate cancer cells via suppressing Nrf2 signaling.
Sigma-Aldrich
Fluorometric Intracellular Ros Kit, sufficient for 200 fluorometric tests (red)
Pricing and availability is not currently available.