Merck
  • Enzymatic treatment of specimens before DNA extraction directly influences molecular detection of infectious agents.
Enzymatic treatment of specimens before DNA extraction directly influences molecular detection of infectious agents.
PloS one (2014-06-18)
Pablo Goldschmidt, Sandrine Degorge, Lilia Merabet, Christine Chaumeil
ABSTRACT
Biological samples, pharmaceuticals or food contain proteins, lipids, polymers, ammoniums and macromolecules that alter the detection of infectious agents by DNA amplification techniques (PCR). Moreover the targeted DNA has to be released from the complex cell walls and the compact nucleoprotein matrixes and cleared from potential inhibitors. The goal of the present work was to assess the efficiency of enzymatic pretreatments on infectious agents to make DNA available for further extraction and amplification. Staphylococcus epidermidis, Streptococcus mitis, Propionibacterium acnes, Escherichia coli, Pseudomonas aeruginosa, Candida albicans, Aspergillus niger and Fusarium solani were mixed with an internal control virus and treated with: 1) proteinase K; 2) lyticase and 3) lyticase followed by proteinase K. DNAs was manually extracted using the QIAmp DNA Mini kit or the MagNA Pure Compact automate. DNA extraction yields and the inhibitors were assessed with a phocid Herpesvirus. Bacterial detection was performed using TaqMan real-time PCR and yeasts and filamentous Fungi with HRM (real-time PCR followed by high-resolution melting analysis). Viral DNA was released, extracted and detected using manual and automatic methods without pre enzymatic treatments. Either the manual or the automatic DNA extraction systems did not meet the sensitivity expectations if enzymatic treatments were not performed before: lyticase for Fungi and Proteinase K for Bacteria. The addition of lyticase and proteinase K did not improve results. For Fungi the detection after lyticase was higher than for Proteinase K, for which melting analysis did not allow fungal specification. Columns and magnetic beads allowed collecting DNA and separate PCR inhibitors. Detection rates cannot be related to DNA-avidity of beads or to elution but to the lack of proteolysis.
Sigma-Aldrich
Lyticase from Arthrobacter luteus, lyophilized powder, ≥200 units/mg solid
Sigma-Aldrich
Lyticase from Arthrobacter luteus, lyophilized powder, ≥2,000 units/mg protein, Protein ≥20 % by biuret