- Comparative study of acid and basic catalysis in microwave assistance of Willgerodt-Kindler reaction for thiobenzamides and derivatives synthesis
Comparative study of acid and basic catalysis in microwave assistance of Willgerodt-Kindler reaction for thiobenzamides and derivatives synthesis
International journal of biological and chemical sciences (2014)
Agnimonhan FH, et al.