Merck
  • Lipopolysaccharide and TNF-alpha produce very similar changes in gene expression in human endothelial cells.
Lipopolysaccharide and TNF-alpha produce very similar changes in gene expression in human endothelial cells.
Journal of vascular research (2006-08-22)
S Magder, J Neculcea, V Neculcea, R Sladek
ABSTRACT
Intracellular signaling pathways regulated by Toll-like receptor 4 (TLR4) and tumor necrosis factor-alpha (TNF-alpha) both activate NFkappaB. This suggests that lipopolysaccharide (LPS) and TNF-alpha should alter transcription of a common set of genes. We tested this hypothesis by treating first passage human umbilical endothelial cells (HUVEC) for 6 h with LPS (50 ng/ml+1 microg/ml CD14) or TNF-alpha (10 ng/ml) and analyzing changes in gene expression by microarray analysis (Affymetrix GeneChips). LPS and TNF-alpha increased expression of 191 common genes and decreased expression of 102 genes. Regulated transcripts encoded for a large number of chemokines, adhesion molecules, procoagulant factors, and molecules that affect cell integrity. Based on the microarray analysis and subsequent confirmation of specific genes by Northern analysis, all 203 genes altered by LPS were altered by TNF-alpha. An additional 17 genes were induced only by TNF-alpha and the expression of 46 was reduced. There were, however, some differences in the kinetics of changes. We also showed that endogenous CD14 was present on these early passage cells and exogenous CD14 was not necessary for most of the LPS response. An autocrine effect from LPS induced expression of TNF-alpha also was ruled out by blocking TNF-alpha with monoclonal antibodies. In conclusion, LPS induces a robust alteration in gene expression in HUVEC that is very similar to that induced by TNF-a. This LPS effect on endothelium could play an important role in the innate immune response.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture