Merck
  • A new rapid and economic liquid chromatographic method for simultaneous determination of meglitinides with metformin: application in the presence of metformin and repaglinide impurities and related compounds
A new rapid and economic liquid chromatographic method for simultaneous determination of meglitinides with metformin: application in the presence of metformin and repaglinide impurities and related compounds
Journal of the Iranian Chemical Society (2018)
El-Zaher AA, et al.