Merck
  • Development of normal phase high-performance liquid chromatographic method for determination of metformin and its related impurities in bulk material
Development of normal phase high-performance liquid chromatographic method for determination of metformin and its related impurities in bulk material
The pharma innovation journal (2017)
Reid IOA